Learn More
Biomatik Corporation Human ASMA (Anti Smooth Muscle Antibody) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57739
Encompass_Preferred
The Human ASMA (Anti Smooth Muscle Antibody) ELISA Kit allows for quantitative determination of ASMA in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 7.813-500 ng/mL. Sensitivity: 4.688 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-0911