Learn More
Biomatik Corporation Rat Ace (Angiotensin-converting enzyme) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57865
Encompass_Preferred
Rat Ace (Angiotensin-converting enzyme) ELISA Kit for detecting Ace in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P47820. Target Synonyms: ACE; CD143; DCP1; Kininase II; ACE1angiotensin converting enzyme; somatic isoform; angiotensin I converting enzyme; peptidyl-dipeptidase A 1; carboxycathepsin; CD143; CD143 antigen; DCP; DCP1angiotensin-converting enzyme; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase I; Kininase II; MVCD3; peptidase P; testicular ECA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1033