Learn More
Biomatik Corporation Rat Ace2 (Angiotensin-converting enzyme 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57924
Encompass_Preferred
Rat Ace2 (Angiotensin-converting enzyme 2) ELISA Kit for detecting Ace2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q5EGZ1. Target Synonyms: Ace2; ACE-related carboxypeptidase;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1085