Learn More
Biomatik Corporation E2 (Estradiol) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57979
Encompass_Preferred
The E2 (Estradiol) ELISA Kit allows for quantitative determination of E2 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 0.703-45 ng/mL. Sensitivity: 0.422 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1128