Learn More
Biomatik Corporation Human Transforming Growth Factor Beta 1 (TGFb1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54966
Encompass_Preferred
Human Transforming Growth Factor Beta 1 (TGFb1) ELISA Kit for detecting TGFb1 in serum, platelet-poor plasma, tissue homogenates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 11.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P01137. Gene ID: 7040. Target Synonyms: TGF-B1; CED; DPD1; LAP; Camurati-Engelmann Disease; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1582