Learn More
Biomatik Corporation Porcine Tumor Necrosis Factor Alpha (TNFa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55004
Encompass_Preferred
Porcine Tumor Necrosis Factor Alpha (TNFa) ELISA Kit for detecting TNFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 62.5-4000pg/ml. Sensitivity: 27.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P23563. Gene ID: 397086. Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2; TNF; Tumor Necrosis Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1598