Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 12 (ADAM12) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU02043
The Human A Disintegrin And Metalloprotease 12 (ADAM12) ELISA Kit allows for quantitative determination of A Disintegrin And Metalloprotease 12 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O43184. Cross Activity: No significant cross-reactivity or interference between A Disintegrin And Metalloprotease 12 and analogues was observed. Restriction: For Research Use Only.