Learn More
Biomatik Corporation Rat Adrenergic Receptor Beta 3 (ADRb3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02173
Encompass_Preferred
Rat Adrenergic Receptor Beta 3 (ADRb3) ELISA Kit for detecting ADRb3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26255. Gene ID: 25645. Target Synonyms: BETA3AR; B3AR; Beta-3 adrenoreceptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1708