Learn More
Biomatik Corporation Advanced Glycation End Product (AGE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02193
Encompass_Preferred
The Advanced Glycation End Product (AGE) ELISA Kit allows for quantitative determination of Advanced Glycation End Product in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 98.8-8,000 ng/mL. Sensitivity: 38.2 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Advanced Glycation End Product and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1719