Learn More
Biomatik Corporation Rat Angiopoietin 2 (ANGPT2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02364
Encompass_Preferred
Rat Angiopoietin 2 (ANGPT2) ELISA Kit for detecting ANGPT2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35462. Gene ID: 89805. Target Synonyms: ANG2; AGPT2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1791