Learn More
Biomatik Corporation Rat Angiotensin I Converting Enzyme (ACE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02397
Encompass_Preferred
Rat Angiotensin I Converting Enzyme (ACE) ELISA Kit for detecting ACE in serum, plasma, cerebrospinal fluid, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47820. Gene ID: 24310. Target Synonyms: CD143; ACE1; DCP1; ACEI; ACE-I; Kininase II; Angiotensin-Converting Enzyme; Peptidyl-Dipeptidase A; Dipeptidyl Carboxypeptidase 1; Angiotensin-converting enzyme, soluble form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1803