Learn More
Biomatik Corporation Cattle Beta-Lactoglobulin (bLg) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU02707
The Cattle Beta-Lactoglobulin (bLg) ELISA Kit allows for quantitative determination of Beta-Lactoglobulin in Serum, Plasma, Tissue homogenates, Cell lysates, milk, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02754. Cross Activity: No significant cross-reactivity or interference between Beta-Lactoglobulin and analogues was observed. Restriction: For Research Use Only.