Learn More
Biomatik Corporation Mouse Cardiotrophin Like Cytokine Factor 1 (CLCF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02956
Encompass_Preferred
Mouse Cardiotrophin Like Cytokine Factor 1 (CLCF1) ELISA Kit for detecting CLCF1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.15pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QZM3. Gene ID: 56708. Target Synonyms: CLC; NR6; BSF3; CISS2; NNT1; Novel Neurotrophin-1; B Cell-Stimulating Factor-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2016