Learn More
Biomatik Corporation Human Chemokine C-C-Motif Ligand 1 (CCL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03092
Encompass_Preferred
The Human Chemokine C-C-Motif Ligand 1 (CCL1) ELISA Kit allows for quantitative determination of Chemokine C-C-Motif Ligand 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 312-5000 pg/mL. Sensitivity: 133 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P22362. Cross Activity: No significant cross-reactivity or interference between Chemokine C-C-Motif Ligand 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2076