Learn More
Biomatik Corporation Human Cluster of Differentiation 24 (CD24) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU03233
The Human Cluster of Differentiation 24 (CD24) ELISA Kit allows for quantitative determination of Cluster of Differentiation 24 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25063. Cross Activity: No significant cross-reactivity or interference between Cluster of Differentiation 24 and analogues was observed. Restriction: For Research Use Only.