Learn More
Biomatik Corporation Dog Complement Component 3a (C3a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03386
Encompass_Preferred
The Dog Complement Component 3a (C3a) ELISA Kit allows for quantitative determination of Complement Component 3a in Serum and Plasma. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.29 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: F1PIX8. Cross Activity: No significant cross-reactivity or interference between Complement Component 3a and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2196