Learn More
Biomatik Corporation Human Enolase, Neuron Specific (NSE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03903
Encompass_Preferred
The Human Enolase, Neuron Specific (NSE) ELISA Kit allows for quantitative determination of Enolase, Neuron Specific in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.231 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09104. Cross Activity: No significant cross-reactivity or interference between Enolase, Neuron Specific and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2398