Learn More
Biomatik Corporation Human Fibroblast Growth Factor 21 (FGF21) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04157
Encompass_Preferred
The Human Fibroblast Growth Factor 21 (FGF21) ELISA Kit allows for quantitative determination of Fibroblast Growth Factor 21 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NSA1. Cross Activity: No significant cross-reactivity or interference between Fibroblast Growth Factor 21 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2496