Learn More
Biomatik Corporation Mouse Laminin Alpha 1 (LAMA1) ELISA Kit
Supplier:  Biomatik Corporation EKU05554
Encompass_Preferred
Mouse Laminin Alpha 1 (LAMA1) ELISA Kit for detecting LAMA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19137. Gene ID: 16772. Target Synonyms: LAM-A1; LAMA; Laminin A; LaMa1; S-laminin subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6181