Learn More
Biomatik Corporation Mouse Galectin 3 (GAL3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04320
Encompass_Preferred
Mouse Galectin 3 (GAL3) ELISA Kit for detecting GAL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8C253. Gene ID: 16854. Target Synonyms: LGALS3; CBP35; GALBP; GALIG; MAC2; 35 kDa lectin; Carbohydrate-binding protein 35; Galactose-specific lectin 3; Galactoside-binding protein; Laminin-binding protein; Lectin L-29. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2554