Learn More
Biomatik Corporation Mouse Glucosidase Alpha, Acid (GaA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04426
Encompass_Preferred
Mouse Glucosidase Alpha, Acid (GaA) ELISA Kit for detecting GaA in serum, plasma, urine, saliva, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70699. Gene ID: 14387. Target Synonyms: LYAG; Acid Alpha-Glucosidase; Lysosomal Alpha-Glucosidase; Pompe Disease Glycogen Storage Disease Type II; Acid Maltase; Aglucosidase Alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2599