Learn More
Biomatik Corporation Mouse Prostate Specific Antigen (PSA) ELISA Kit
Supplier:  Biomatik Corporation EKU05439
Encompass_Preferred
Mouse Prostate Specific Antigen (PSA) ELISA Kit for detecting PSA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00756. Gene ID: 18050. Target Synonyms: KLK3; APS; KLK2A1; HK3; KLKB1; Seminin; Prostate Specific Antigen; Kallikrein-Related Peptidase 3; Semenogelase, g-Seminoprotein; P-30 Antigen; Plasma kallikrein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6161