Learn More
Biomatik Corporation Rat Growth Differentiation Factor 11 (GDF11) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04587
Encompass_Preferred
Rat Growth Differentiation Factor 11 (GDF11) ELISA Kit for detecting GDF11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z217. Gene ID: 29454. Target Synonyms: BMP11; Bone morphogenetic protein 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2660