Learn More
Biomatik Corporation Human Protein Kinase, AMP Activated Alpha 1 (AMPK Alpha 1) ELISA Kit
Supplier:  Biomatik Corporation EKU06929
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q13131. Target Synonyms: PRKAa1; AMPK; AMPKa1; PKA; ACACA kinase; 5'-AMP-Activated Protein Kinase Catalytic Subunit Alpha-1; Acetyl-CoA carboxylase kinase; Hydroxymethylglutaryl-CoA reductase kinase. Restriction: For Research Use Only.
Catalog No. 50-228-6344