Learn More
Biomatik Corporation Human Growth Hormone Releasing Hormone (GHRH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04617
Encompass_Preferred
The Human Growth Hormone Releasing Hormone (GHRH) ELISA Kit allows for quantitative determination of Growth Hormone Releasing Hormone in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 4.88 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01286. Cross Activity: No significant cross-reactivity or interference between Growth Hormone Releasing Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2673