Learn More
Biomatik Corporation Human Rh Blood Group, D Antigen (RHD) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU10646
Encompass_Preferred
Human Rh Blood Group, D Antigen (RHD) Anti-antigen ELISA Kit for detecting RHD in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.27ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: Q02161. Target Synonyms: CD240D; Rh30a; Rh4; RhPI; RhII; DIIIc; Rhesus D antigen; Rh polypeptide 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6729