Learn More
Biomatik Corporation Mouse Heat Shock 70kDa Protein 8 (HSPA8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04664
Encompass_Preferred
Mouse Heat Shock 70kDa Protein 8 (HSPA8) ELISA Kit for detecting HSPA8 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63017. Gene ID: 15481. Target Synonyms: LAP1; HSC54; HSC70; HSC71; HSP71; HSPA10; NIP71; HSP73; Heat shock cognate 71 kDa protein; Lipopolysaccharide-associated protein 1; LPS-associated protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2694