Learn More
Biomatik Corporation Human Heat Shock Protein 47 (HSP47) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04668
Encompass_Preferred
The Human Heat Shock Protein 47 (HSP47) ELISA Kit allows for quantitative determination of Heat Shock Protein 47 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8,000 pg/mL. Sensitivity: 46 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P50454. Cross Activity: No significant cross-reactivity or interference between Heat Shock Protein 47 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2697