Learn More
Biomatik Corporation Human Collagen Type Iii, Col Iii Elisa Kit
Supplier:  Biomatik Corporation EKC33225
Encompass_Preferred
Human Collagen Type Iii, Col Iii Elisa Kit for detecting Collagen Type III, Col III in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800ng/ml. Sensitivity: 3.12ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: Collagen Type 3; Col 3; Collagen Type III; Col III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7371