Learn More
Biomatik Corporation Rat dynorphin,Dyn ELISA Kit
Supplier:  Biomatik Corporation EKC40971
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 2.35 pg/mL-150 pg/mL. Sensitivity: 0.58 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7807