Learn More
Biomatik Corporation Mouse Insulin Like Growth Factor Binding Protein 7 (IGFBP7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05009
Encompass_Preferred
Mouse Insulin Like Growth Factor Binding Protein 7 (IGFBP7) ELISA Kit for detecting IGFBP7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 28.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61581. Gene ID: 29817. Target Synonyms: FSTL2; IGFBP-7v; MAC25; PSF; TAF; IGFBP-rP1; MAC25 protein; Prostacyclin-stimulating factor; PGI2-stimulating factor; Tumor-derived adhesion factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2828