Learn More
Biomatik Corporation Gallus Interferon Beta (IFNb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05090
Encompass_Preferred
The Gallus Interferon Beta (IFNb) ELISA Kit allows for quantitative determination interferon Beta in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 3.12 -200 pg/mL. Sensitivity: 1.22 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q90873. Cross Activity: No significant cross-reactivity or interference between Interferon Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2867