Learn More
Biomatik Corporation Cattle Interferon Gamma (IFNg) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU05096
The Cattle Interferon Gamma (IFNg) ELISA Kit allows for quantitative determination interferon Gamma in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07353. Cross Activity: No significant cross-reactivity or interference between Interferon Gamma and analogues was observed. Restriction: For Research Use Only.