Learn More
Biomatik Corporation Human Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit
Supplier:  Biomatik Corporation EKU05156
Encompass_Preferred
Sample Type: Serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Human. Detection Range: 31.2-2,000pg/mL. Sensitivity: 12.9pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P18510. Target Synonyms: IL1RN; IL1-RA; ICIL1RA; IL1ra3; IL1F3; IRAP;IL-1RA; Interleukin-1 Family Member 3; IL1 inhibitor; Anakinra. Restriction: For Research Use Only.
Catalog No. 50-228-6121