Learn More
Biomatik Corporation Mouse Plasminogen Activator, Urokinase (uPA) ELISA Kit
Supplier:  Biomatik Corporation EKU06672
Encompass_Preferred
Mouse Plasminogen Activator, Urokinase (uPA) ELISA Kit for detecting uPA in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06869. Gene ID: 18792. Target Synonyms: PLAU; ATF; URK; UK; UP-A; Abbokinase; Urokinase-Type Plasminogen Activator. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6315