Learn More
Biomatik Corporation Rat Cyclooxygenase-2 (COX 2) ELISA Kit
Supplier:  Biomatik Corporation EKU06863
Encompass_Preferred
Rat Cyclooxygenase-2 (COX 2) ELISA Kit for detecting COX 2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35355. Gene ID: 29527. Target Synonyms: PGG/HS; PGHS2; PHS2; HCox2; PTGS2; Prostaglandin Endoperoxide Synthase 2; Prostaglandin G/H Synthase And Cyclooxygenase; Prostaglandin H2 synthase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6336