Learn More
Biomatik Corporation Human Interleukin 17 Receptor A (IL17RA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05240
Encompass_Preferred
The Human Interleukin 17 Receptor A (IL17RA) ELISA Kit allows for quantitative determination interleukin 17 Receptor A in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 30 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96F46. Cross Activity: No significant cross-reactivity or interference between Interleukin 17 Receptor A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2951