Learn More
Biomatik Corporation Human Salusin Alpha (SALa) ELISA Kit
Supplier:  Biomatik Corporation EKU07190
Encompass_Preferred
Human Salusin Alpha (SALa) ELISA Kit for detecting SALa in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.47-200pg/ml. Sensitivity: 0.93pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q8N2E6. Gene ID: 27433. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6367