Learn More
Biomatik Corporation Tetracycline (TTC) ELISA Kit
Supplier:  Biomatik Corporation EKU11588
Encompass_Preferred
Tetracycline (TTC) ELISA Kit for detecting TTC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 123.5-10,000pg/ml. Sensitivity: 50.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Sumycin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6961