Learn More
Biomatik Corporation Human Interleukin 4 Induced Protein 1 (IL4I1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05341
Encompass_Preferred
The Human Interleukin 4 Induced Protein 1 (IL4I1) ELISA Kit allows for quantitative determination interleukin 4 Induced Protein 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96RQ9. Cross Activity: No significant cross-reactivity or interference between Interleukin 4 Induced Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3013