Learn More
Biomatik Corporation Human Angiopoietin 2, Ang-2 Elisa Kit
Supplier:  Biomatik Corporation EKC32574
Encompass_Preferred
Human Angiopoietin 2, Ang-2 Elisa Kit for detecting Angiopoietin 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 39-2500pg/ml. Sensitivity: 9.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O15123; ANGPT2. Target Synonyms: AGPT 2; Agpt2; ANG 2; ANG-2; ANG2; Angiopoietin 2a; Angiopoietin 2B; Angiopoietin-2; Angiopoietin2; ANGP2_HUMAN; ANGPT 2; Angpt2; Tie2 ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7312