Learn More
Biomatik Corporation Mouse Keratin 18 (KRT18) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05467
Encompass_Preferred
Mouse Cytokeratin 18 (CK18) ELISA Kit for detecting CK18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05784. Gene ID: 16668. Target Synonyms: KRT18; CYK18; K18; Keratin 18; Cell proliferation-inducing gene 46 protein; Keratin, type I cytoskeletal 18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3070