Learn More
Biomatik Corporation Pig Substance P(SP)ELISA Kit
Supplier:  Biomatik Corporation EKC38337
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 0.25 ng/mL-16 ng/mL. Sensitivity: 0.06 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7639