Learn More
Biomatik Corporation Human Kisspeptin 1 (KISS1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05504
Encompass_Preferred
The Human Kisspeptin 1 (KISS1) ELISA Kit allows for quantitative determination of Kisspeptin 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 13.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q15726. Cross Activity: No significant cross-reactivity or interference between Kisspeptin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3084