Learn More
Biomatik Corporation Human Leukotriene A4 Hydrolase (LTA4H) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05630
Encompass_Preferred
The Human Leukotriene A4 Hydrolase (LTA4H) ELISA Kit allows for quantitative determination of Leukotriene A4 Hydrolase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.128 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09960. Cross Activity: No significant cross-reactivity or interference between Leukotriene A4 Hydrolase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3130