Learn More
Biomatik Corporation Human Lipopolysaccharide Binding Protein (LBP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05662
Encompass_Preferred
The Human Lipopolysaccharide Binding Protein (LBP) ELISA Kit allows for quantitative determination of Lipopolysaccharide Binding Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.29 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P18428. Cross Activity: No significant cross-reactivity or interference between Lipopolysaccharide Binding Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3145