Learn More
Biomatik Corporation Human Methylenetetrahydrofolate Reductase (MTHFR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05939
Encompass_Preferred
The Human Methylenetetrahydrofolate Reductase (MTHFR) ELISA Kit allows for quantitative determination of Methylenetetrahydrofolate Reductase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 23.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P42898. Cross Activity: No significant cross-reactivity or interference between Methylenetetrahydrofolate Reductase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3263