Learn More
Biomatik Corporation Mouse Neurofilament, Light Polypeptide (NEFL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06216
Encompass_Preferred
Mouse Neurofilament, Light Polypeptide (NEFL) ELISA Kit for detecting NEFL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08551. Gene ID: 18039. Target Synonyms: CMT1F; CMT2E; NF-L; NF68; NFL; 68 kDa neurofilament protein; Neurofilament triplet L protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3353