Learn More
Biomatik Corporation Human Parathyroid Hormone Receptor 1 (PTHR1) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU06486
The Human Parathyroid Hormone Receptor 1 (PTHR1) ELISA Kit allows for quantitative determination of Parathyroid Hormone Receptor 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.127 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q03431. Cross Activity: No significant cross-reactivity or interference between Parathyroid Hormone Receptor 1 and analogues was observed. Restriction: For Research Use Only.