Learn More
Biomatik Corporation Human N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) CLIA Kit
Supplier:  Biomatik Corporation EKU09463
Encompass_Preferred
Human N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) CLIA Kit for detecting NT-ProBNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.43-2,500pg/ml. Sensitivity: 1.33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P16860. Gene ID: 4879. Target Synonyms: NT-Pro-BNP; ; N-BNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6618